CAS RN 1335106-03-0
IUPAC Nombre:
N-(5-{[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]sulfamoyl}-4-methyl-1,3-thiazol-2-yl)acetamide
Sinónimos:
N-(5-{[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]sulfamoyl}-4-methyl-1,3-thiazol-2-yl)acetamide
Peso molecular (g/mol):
477.4
Fórmula molecular:
C15H13F6N3O4S2
InChi Key:
OZBSSKGBKHOLGA-UHFFFAOYSA-N
SMILES:
CC(=O)NC1=NC(C)=C(S1)S(=O)(=O)NC1=CC=C(C=C1)C(O)(C(F)(F)F)C(F)(F)F
1
–
1
de
1
resultados
1
SR1001, MedChemExpress
MedChemExpress SR1001 is a selective RORα and RORγt inverse agonist with Kis 172 and 111 nM, respectively.