CAS RN 13299-99-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MAT2A inhibitor 2, MedChemExpress
MedChemExpress MAT2A inhibitor 2 is a methionine adenosyltransferase 2A (MAT2A) inhibitor.