CAS RN 1329610-82-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
O,O-Dimetil Ditiofosfato-13C2 Sal de Amonio, TRC
CAS: 1329610-82-3 Fórmula molecular: 13C2H10NO2PS2 Peso molecular (g/mol): 177.2 Sinónimo: Phosphorodithioic acid O,O-di(methyl-13C) ester ammonium salt,[13C]Methyl ammonium phosphorodithioate,Ammonium O,O-di(methyl-13C) dithiophosphate,O,O-Di(methyl-13C) dithiophosphoric acid ammonium salt,O,O-Di(methyl-13C) ammonium dithiophosphate,O,O-Dimethyl ammonium phosphorodithioate-13C2,O,O-Dimethyl phosphorodithioic acid-13C2 ammonium salt,O,O-Dimethylphosphorodithioic acid 13C2 ammonium salt SMILES: [NH4+].[13CH3]OP(=S)([S-])O[13CH3]