CAS RN 1324003-64-6
IUPAC Nombre:
3-(4-methoxypyridine-3-carbonyl)-1-[2-(trifluoromethyl)-[1,1'-biphenyl]-4-yl]urea
Sinónimos:
3-(4-methoxypyridine-3-carbonyl)-1-[2-(trifluoromethyl)-[1,1'-biphenyl]-4-yl]urea
Peso molecular (g/mol):
415.37
Fórmula molecular:
C21H16F3N3O3
InChi Key:
MLDQTQOMWDNTNN-UHFFFAOYSA-N
SMILES:
COC1=C(C=NC=C1)C(=O)NC(=O)NC1=CC(=C(C=C1)C1=CC=CC=C1)C(F)(F)F
1
–
1
de
1
resultados
1
S1P1 agonist III, MedChemExpress
MedChemExpress S1P1 Agonist III is a potent and orally active S1P1 agonist with EC50 of 18 nM; no activity on S1P3.