CAS RN 1312445-63-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Buparlisib Hydrochloride, MedChemExpress
MedChemExpress Buparlisib Hydrochloride (BKM120 Hydrochloride) is a pan-class I PI3K inhibitor, with IC50 of 52 nM/166 nM/116 nM/262 nM for p110α/p110β/p110δ/p110γ, respectively.