CAS RN 1303607-07-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MI-773, MedChemExpress
MedChemExpress MI-773 is a potent MDM2-p53 protein‐protein interaction (PPI) inhibitor with high binding affinity against MDM2 (Kd=8.2 nM). MI-773 has antitumor activity.