CAS RN 1273323-67-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
YLF-466D, MedChemExpress
MedChemExpress YLF-466D is a newly developed AMPK activator, which inhibits platelet aggregation.