CAS RN 127274-91-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ DiD perchlorate
Lipophilic fluorescent reagent; shown to stain HSCs
2
MedChemExpress DiD perchlorate
MedChemExpress DiD perchlorate is a far-red fluorescent lipophilic cyanine dye. DiD perchlorate can rapidly and stably integrate into the phospholipid cell membrane. DiD perchlorate is used to cells tracking.
3