CAS RN 1255517-76-0
IUPAC Nombre:
2-{[4-(5-ethylpyrimidin-4-yl)piperazin-1-yl]methyl}-6-(trifluoromethyl)-1H-1,3-benzodiazole
Sinónimos:
2-{[4-(5-ethylpyrimidin-4-yl)piperazin-1-yl]methyl}-5-(trifluoromethyl)-3H-1,3-benzodiazole
Peso molecular (g/mol):
390.41
Fórmula molecular:
C19H21F3N6
InChi Key:
FBLPQCAQRNSVHB-UHFFFAOYSA-N
SMILES:
CCC1=CN=CN=C1N1CCN(CC2=NC3=CC=C(C=C3N2)C(F)(F)F)CC1
1
–
2
de
2
resultados
1
Tocris Bioscience™ PF 4708671
S6K1 inhibitor
2
PF-4708671, MedChemExpress
MedChemExpress PF-4708671 is a potent cell-permeable S6K1 inhibitor with a Ki of 20 nM and IC50 of 160 nM.