CAS RN 1253186-49-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GSK-5498A, MedChemExpress
MedChemExpress GSK-5498A is a selective small molecule blocker of CARC (IC50, 1 μM); inhibits mediator release from mast cells, and pro-inflammatory cytokine release from T-cells in a variety of species.