CAS RN 1246089-97-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
(Rac)-Pyrotinib, MedChemExpress
MedChemExpress (Rac)-Pyrotinib ((Rac)-SHR-1258) is the racemate of Pyrotinib. Pyrotinib is a potent and selective EGFR/HER2 dual inhibitor.