CAS RN 1234563-16-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Epetraborole hydrochloride, MedChemExpress
MedChemExpress Epetraborole hydrochloride is a novel leucyl-tRNA synthetase (LeuRS) inhibitor, which inhibits protein synthesis by binding to the terminal adenosine ribose (A76) of leucyl-tRNA synthetase.