Learn More
Epetraborole hydrochloride, MedChemExpress
Código de producto. 16497917
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
1 mg
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
1 miligramo
10 miligramos
100 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16497917 5 mg 5 miligramos
16481259 1 mg 1 miligramo
16475886 100 mg 100 miligramos
16445198 10 mg 10 miligramos
16441417 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16497917 Proveedor Medchem Express N.º de proveedor HY12479A
MedChemExpress Epetraborole hydrochloride is a novel leucyl-tRNA synthetase (LeuRS) inhibitor, which inhibits protein synthesis by binding to the terminal adenosine ribose (A76) of leucyl-tRNA synthetase.
Epetraborole hydrochloride is a novel leucyl-tRNA synthetase (LeuRS) inhibitor, which inhibits protein synthesis by binding to the terminal adenosine ribose (A76) of leucyl-tRNA synthetase.
Especificaciones
Nombre del producto químico o material Epetraborole hydrochloride
CAS 1234563-16-6
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C11H17BClNO4
Cantidad 5 mg
Sinónimo GSK2251052 hydrochloride
Información de solubilidad DMSO : 200 mg/mL (731.21 mM; Need ultrasonic) ∣H2O : ≥ 28 mg/mL (102.37 mM)
SMILES OCCCOC1=C(B(O)O[C@@H]2CN)C2=CC=C1.[H]Cl
Peso molecular (g/mol) 273.52
Formula Weight (peso de la fórmula) 273.52
Porcentaje de pureza 99.25%
Grado Research
Forma física Powder
Para utilizar con (aplicación) COVID-19-immunoregulation