CAS RN 1228013-30-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Onatasertib, MedChemExpress
MedChemExpress Onatasertib (CC-223) is a potent, selective, and orally bioavailable inhibitor of mTOR kinase, with an IC50 value for mTOR kinase of 16 nM. Onatasertib inhibits both mTORC1 and mTORC2.