CAS RN 1227948-02-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BI-4464, MedChemExpress
MedChemExpress BI-4464 is a highly selective ATP competitive inhibitor of PTK2/FAK, with an IC50 of 17 nM. A PTK2 ligand for PROTAC.