CAS RN 1226781-44-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Omarigliptin, MedChemExpress
MedChemExpress Omarigliptin(MK-3102) is a potent, selective and long-acting DPP-4 inhibitor with IC50 of 1.6 nM; highly selective over all proteases tested (IC50 > 67 (mu)M).