CAS RN 1219168-18-9
IUPAC Nombre:
4-(6-{4-[2-(piperidin-1-yl)ethoxy]phenyl}pyrazolo[1,5-a]pyrimidin-3-yl)pyridine dihydrochloride
Sinónimos:
4-(6-{4-[2-(piperidin-1-yl)ethoxy]phenyl}pyrazolo[1,5-a]pyrimidin-3-yl)pyridine dihydrochloride
Peso molecular (g/mol):
472.41
Fórmula molecular:
C24H27Cl2N5O
InChi Key:
RJDVIJJQKMGPMV-UHFFFAOYSA-N
SMILES:
Cl.Cl.C(CN1CCCCC1)OC1=CC=C(C=C1)C1=CN2N=CC(=C2N=C1)C1=CC=NC=C1
1
–
2
de
2
resultados
1
Tocris Bioscience™ Dorsomorphin dihydrochloride
CAS: 1219168-18-9 Fórmula molecular: C24H27Cl2N5O
2
Dorsomorphin dihydrochloride, MedChemExpress
MedChemExpress Dorsomorphin dihydrochloride (BML-275 dihydrochloride; Compound C dihydrochloride) is a potent, selective and ATP-competitive AMPK inhibitor, with a Ki of 109 nM. Dorsomorphin dihydrochloride inhibits BMP pathway by targeting the type I receptors ALK2, ALK3, and ALK6. Dorsomorphin dihydrochloride induces autophagy.