CAS RN 1216744-19-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
GSK 4112, TRC
CAS: 1216744-19-2 Fórmula molecular: C18H21ClN2O4S Peso molecular (g/mol): 396.89 Sinónimo: GSK 4112 SMILES: CC(C)(C)OC(=O)CN(Cc1ccc(Cl)cc1)Cc2ccc(s2)[N+](=O)[O-]
2
GSK4112, MedChemExpress
MedChemExpress GSK4112 is a Rev-erbα agonist with EC50 of 0.4 μM, also is a small molecule chemical probe for the cell biology of the nuclear heme receptor Rev-erbα.