CAS RN 121524-09-2
IUPAC Nombre:
hydrogen ethyl 2-{[(7S)-7-{[(2R)-2-(3-chlorophenyl)-2-hydroxyethyl]amino}-5,6,7,8-tetrahydronaphthalen-2-yl]oxy}acetate chloride
Sinónimos:
amibegron
Peso molecular (g/mol):
440.36
Fórmula molecular:
C22H27Cl2NO4
InChi Key:
NQIZCDQCNYCVAS-RQBPZYBGSA-N
SMILES:
[H+].[Cl-].CCOC(=O)COC1=CC=C2CC[C@@H](CC2=C1)NC[C@H](O)C1=CC=CC(Cl)=C1
1
–
2
de
2
resultados
1
Tocris Bioscience™ SR 58611A hydrochloride
CAS: 121524-09-2 Fórmula molecular: C22H27Cl2NO4
2
Amibegron hydrochloride, MedChemExpress
MedChemExpress Amibegron hydrochloride is a selective β3-adrenoceptor agonist, with an EC50 of 3.5 nM for β-adrenoceptor in rat colon; Amibegron hydrochloride has anxiolytic and antidepressant activity.