CAS RN 1210945-69-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ EG 00229 trifluoroacetate
Neuropilin 1 (NRP1) receptor antagonist; inhibits VEGFA binding to NRP1
2
EG00229, MedChemExpress
MedChemExpress EG00229 is a neuropilin 1 (NRP1) receptor antagonist. EG00229 selectively inhibits VEGF-A binding to NRP1 b1 domain with an IC50 of 3 μM, but has no effect on VEGFA binding to VEGFR-1 and VEGFR-2.