CAS RN 1210004-12-8
IUPAC Nombre:
4-nitrophenyl 4-[(3-phenoxyphenyl)methyl]piperazine-1-carboxylate
Sinónimos:
4-nitrophenyl 4-[(3-phenoxyphenyl)methyl]piperazine-1-carboxylate
Peso molecular (g/mol):
433.46
Fórmula molecular:
C24H23N3O5
InChi Key:
QNYRAEKLMNDRFY-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(OC(=O)N2CCN(CC3=CC(OC4=CC=CC=C4)=CC=C3)CC2)C=C1
1
–
2
de
2
resultados
1
Tocris Bioscience™ JZL 195
CAS: 1210004-12-8 Fórmula molecular: C24H23N3O5
2
JZL195, MedChemExpress
MedChemExpress JZL195 is a selective and efficacious dual fatty acid amide hydrolase (FAAH) and monoacylglycerol lipase (MAGL) inhibitor with IC50s of 2 and 4 nM, respectively.