CAS RN 1208229-58-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tepilamide fumarate, MedChemExpress
MedChemExpress Tepilamide fumarate (XP-23829) is an oral fumaric acid ester, acts as a prodrug of monomethyl fumarate, and is used in the research of moderate to severe chronic plaque psoriasis.