CAS RN 1204707-73-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SRT3190, MedChemExpress
MedChemExpress SRT3190 is an antagonist of CXCR2, used in the research of chemokine mediated diseases.