CAS RN 1203680-76-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ AS 1949490
SH2 domain-containing inositol 5'-phosphatase 2 (SHIP2) inhibitor
2
AS1949490, MedChemExpress
MedChemExpress AS1949490 is a potent and selective SHIP-2 (SH2 domain-containing inositol 5′ phosphatase 2) inhibitor, with an IC50 of 620 nM. AS1949490 activated glucose metabolism via up-regulation of GLUT1 gene in L6 myotubes.