CAS RN 120241-79-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
3-TYP, MedChemExpress
MedChemExpress 3-TYP is a selective SIRT3 inhibitor, with an IC50 of 16 nM, more potent over SIRT1 (IC50=88 nM), SIRT2 (IC50=92 nM).