CAS RN 1194506-26-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Fruquintinib, MedChemExpress
MedChemExpress Fruquintinib (HMPL-013) is a highly potent and selective VEGFR 1/2/3 inhibitor with IC50s of 33, 0.35, and 35 nM, respectively.