CAS RN 119418-04-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ Galanin (1-30) (human)
Endogenous galanin receptor agonist
2
MedChemExpress Galanin (1-30), human
MedChemExpress Galanin (1-30), human is a 30-amino acid neuropeptide, and acts as an agonist of GalR1 and GalR2 receptors, with Kis of both 1 nM.
3
MedChemExpress Galanin (1-30), human
MedChemExpress Galanin (1-30), human is a 30-amino acid neuropeptide, and acts as an agonist of GalR1 and GalR2 receptors, with Kis of both 1 nM.