CAS RN 1193383-09-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
JNJ 42041935, TRC
CAS: 1193383-09-3 Fórmula molecular: C12 H6 Cl F3 N4 O3 Peso molecular (g/mol): 346.65 Sinónimo: 1-[6-Chloro-5-(trifluoromethoxy)-1H-benzimidazol-2-yl]-1H-pyrazole-4-carboxylic Acid,1-(6-Chloro-5-(trifluoromethoxy)-1H-benzo[d]imidazol-2-yl)-1H-pyrazole-4-carboxylic Acid Nombre IUPAC: 1-[6-cloro-5-(trifluorometaxi)-1H-benzimidazol-2-il]pirazol-4-carboxílico ácido SMILES: OC(=O)c1cnn(c1)c2nc3cc(OC(F)(F)F)c(Cl)cc3[nH]2
2
JNJ-42041935, MedChemExpress
MedChemExpress JNJ-42041935 is a potent, competitive and selective inhibitor of prolyl hydroxylase PHD; inhibits PHD1, PHD2, and PHD3 with pKi values of 7.91±0.04, 7.29 ±0.05, and 7.65±0.09, respectively.