CAS RN 1192189-69-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
LX7101, MedChemExpress
MedChemExpress LX7101 is a potent inhibitor of LIMK and ROCK2 with IC50 values of 24, 1.6 and 10 nM for LIMK1, LIMK2 and ROCK2, respectively; also inhibits PKA with an IC50 less than 1 nM.