CAS RN 1188265-43-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Midodrine D6 hydrochloride, MedChemExpress
MedChemExpress Midodrine D6 hydrochloride is deuterium labeled Midodrine, which is a vasopressor/antihypotensive agent.