CAS RN 1184940-47-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
EMD638683 R-Form, MedChemExpress
MedChemExpress EMD638683 R-Form is the R-form of EMD638683. EMD638683 is a highly selective SGK1 inhibitor with IC50 of 3 μM.