CAS RN 118409-62-4
IUPAC Nombre:
2-[(3-hydroxy-4-nitrophenyl)methylidene]propanedinitrile
Sinónimos:
2-[(3-hydroxy-4-nitrophenyl)methylidene]propanedinitrile
Peso molecular (g/mol):
215.17
Fórmula molecular:
C10H5N3O3
InChi Key:
DUQADSPERJRQBW-UHFFFAOYSA-N
SMILES:
OC1=CC(C=C(C#N)C#N)=CC=C1[N+]([O-])=O
1
–
1
de
1
resultados
1
AG126, MedChemExpress
MedChemExpress AG126 is a tyrosine kinase inhibitor which can prevent the activation of mitogen-activated protein kinase p42MAPK (ERK2).