CAS RN 118-08-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Hydrastine, MedChemExpress
MedChemExpress Hydrastine is a natural alkaloid which is present in Hydrastis canadensis and other plants of the ranunculaceae family.