CAS RN 117047-07-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
3'-Methoxypuerarin, MedChemExpress
MedChemExpress 3'-Methoxypuerarin (3'-MOP) is an isoflavone extracted from radix puerariae that shows neuron protection activity.