CAS RN 1166227-08-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ A66
Potent and selective PI 3-kinase p110α inhibitor
2
A66, MedChemExpress
MedChemExpress A66 is a highly specific and selective p110α inhibitor with an IC50 of 32 nM.
3