CAS RN 1161921-82-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BAY-85-8501, MedChemExpress
MedChemExpress BAY-85-8501 is a selective, reversible and potent inhibitor of Human Neutrophil Elastase (HNE), with an IC50 of 65 pM.