CAS RN 1116743-46-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SAR125844, MedChemExpress
MedChemExpress SAR125844 is a potent, highly selective, reversible and ATP-competitive MET receptor tyrosine kinase (RTK) inhibitor, with an IC50 of 4.2 nM. Shows inhibition of MET autophosphorylation in cell-based assays.