CAS RN 1108743-60-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Entrectinib, MedChemExpress
MedChemExpress Entrectinib (NMS-E628) is a potent, orally available, and CNS-active pan-Trk, ROS1, and ALK inhibitor. Entrectinib inhibits TrkA, TrkB, TrkC, ROS1 and ALK with IC50 values of 1, 3, 5, 12 and 7 nM, respectively. Antitumor activity.