CAS RN 1094873-14-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
JNJ-31020028, MedChemExpress
MedChemExpress JNJ-31020028 is a selective brain penetrant antagonist of neuropeptide Y2 receptor with high affinity (pIC50=8.07, human; pIC50=8.22 rat); >100-fold selective versus human Y1/Y4/Y5 receptors.