CAS RN 1094069-99-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SCH-1473759, MedChemExpress
MedChemExpress SCH-1473759 is an aurora inhibitor with IC50s of 4 and 13 nM for aurora A and B, respectively.