Learn More
Tocris Bioscience™ ZM 447439
Descripción
Novel, selective ATP-competitive inhibitor of Aurora B kinase in vitro (IC50 values are 50, 250 and 1000 nM for Aurora B, C and A kinases respectively). Selective over a range of other kinases including cdk1 and PLK1 (IC50 > 10 μM). Inhibits cell division and displays selective toxicity towards proliferating tumor cells versus non-dividing cells.
Especificaciones
Especificaciones
| Nombre del producto químico o material | ZM 447439 |
| CAS | 331771-20-1 |
| Cantidad | 10 mg |
| Fórmula molecular | C29H31N5O4 |
| Formula Weight (peso de la fórmula) | 513.59 |
| Pureza | >99% |
| Peso molecular | 513.598 |