Learn More
ZIP4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Código de producto. 16870185 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16870185 20 μL 20 microlitros
16860185 150 μL 150 microlitros
2 options
Código de producto. 16870185
Marca: Proteintech 206251AP20UL
Rabbit Polyclonal Antibody
The zinc transporter ZIP4, also known as SLC39A4, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP4 was initially identified as zinc-uptake transporter in the intestine and as a candidate gene for acrodermatitis enteropathica (AE), a rare inherited defect in the absorption of dietary zinc. Similar to other ZIP family members, aberrant expression of ZIP4 has been linked to cancer, specifically the progression and aggressiveness of pancreatic cancer. ZIP4 expression has also been shown to be activated in hepatocellular carcinomas, acting to repress apoptosis, enhance cell cycle and increase migration, suggesting ZIP4 may play a role in multiple types of cancers.
Especificaciones
Antígeno ZIP4
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.51 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica SLC39A4
N.º de referencia del gen Q6P5W5, Q78IQ7
Alias de gen AEZ, SLC39A4, Zinc transporter ZIP4, ZIP 4, ZIP4, Zrt and Irt like protein 4
Símbolos de los genes SLC39A4
Especie del huésped Rabbit
Inmunógeno ZIP4 Fusion Protein Ag14524
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 55630, 72027
Especies diana Mouse, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG