Learn More
Zinc Pyrithione, MedChemExpress
Código de producto. 16442239
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
1 g
5 g
Tamaño de la unidad:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16442239 1 g 1 g
16431008 5 g 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16442239 Proveedor Medchem Express N.º de proveedor HYB0572
MedChemExpress Zinc Pyrithione is an antifungal and antibacterial agent disrupting membrane transport by blocking the proton pump. Zinc Pyrithione is also a copper ionophore that delivers copper into cells and is a useful tool for studying cuproptosis.
Zinc Pyrithione is an antifungal and antibacterial agent disrupting membrane transport by blocking the proton pump. Zinc Pyrithione is also a copper ionophore that delivers copper into cells and is a useful tool for studying cuproptosis.
Especificaciones
Nombre del producto químico o material Zinc Pyrithione
Forma física Solid
CAS 13463-41-7
Color Off-White
Cantidad 1 g
Notas de grado de pureza Research
Fórmula molecular C10H8N2O2S2Zn
Para utilizar con (aplicación) COVID-19-anti-virus
Información de solubilidad DMSO : 33.33 mg/mL (104.91 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.31 mM; Need ultrasonic)
SMILES N12[O-][Zn+2]3([S]=C1C=CC=C2)[O-]N(C=CC=C4)C4=[S]3
Peso molecular (g/mol) 317.69
Formula Weight (peso de la fórmula) 317.69
Porcentaje de pureza 98.0%