missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZC3H13 Polyclonal Antibody

Descripción
Antibody detects endogenous levels of total ZC3H13.
The zinc finger CCCH domain-containing protein 13 (ZC3H13) is a 1668 amino acid protein that contains one C3H1-type zinc finger. ZC3H13 is phosphorylated upon DNA damage, most likely by ATM or ATR. Two isoforms of ZC3H13 exists as a result of alternative splicing events. The gene encoding ZC3H13 maps to chromosome 13, which contains around 114 million base pairs and 400 genes. Key tumor suppressor genes on chromosome 13 include the breast cancer susceptibility gene, BRCA2, and the RB1 (retinoblastoma) gene. As with most chromosomes, polysomy of part or all of chromosome 13 is deleterious to development and decreases the odds of survival. Trisomy 13, also known as Patau syndrome, is quite deadly and the few who survive past one year suffer from permanent neurologic defects, difficulty eating and vulnerability to serious respiratory infections.
Especificaciones
Especificaciones
| Antígeno | ZC3H13 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | ZC3H13 |
| N.º de referencia del gen | E9Q784, Q5T200 |
| Alias de gen | 2600010B19Rik; 3110050K21Rik; 4930570G11Rik; C87618; KIAA0853; RGD1307729; ZC3H13; Zinc finger CCCH domain-containing protein 13; zinc finger CCCH type containing 13; zinc finger CCCH-type containing 13 |
| Símbolos de los genes | ZC3H13 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?