Learn More
Zaltoprofen, MedChemExpress
Código de producto. 16448558
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16448558 10 mg 10 miligramos
16438396 100 mg 100 miligramos
16434427 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16448558 Proveedor Medchem Express N.º de proveedor HYB0619
MedChemExpress Zaltoprofen (CN100), a non-steroidal anti-inflammatory drug (NSAID), is a preferential and orally active COX-2 inhibitor, with IC50s of 1.3 and 0.34 μM for COX-1 and COX-2, respectively. Zaltoprofen exhibits powerful anti-inflammatory effects as well as an analgesic action on inflammatory pain.
Zaltoprofen (CN100), a non-steroidal anti-inflammatory drug (NSAID), is a preferential and orally active COX-2 inhibitor, with IC50s of 1.3 and 0.34 μM for COX-1 and COX-2, respectively. Zaltoprofen exhibits powerful anti-inflammatory effects as well as an analgesic action on inflammatory pain.
Especificaciones
Nombre del producto químico o material Zaltoprofen
Forma física Solid
CAS 74711-43-6
Color Off-White
Cantidad 10 mg
Notas de grado de pureza Research
Fórmula molecular C17H14O3S
Para utilizar con (aplicación) Neuroscience-Neuromodulation
Sinónimo CN100
Información de solubilidad DMSO : ≥ 100 mg/mL (335.17 mM)
SMILES CC(C1=CC=C(SC2=C3C=CC=C2)C(CC3=O)=C1)C(O)=O
Peso molecular (g/mol) 298.36
Formula Weight (peso de la fórmula) 298.36
Porcentaje de pureza 98.0%