Learn More
MedChemExpress Z-IETD-FMK
Product Code. 16416057
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
Unit Size:
5 miligramos
Product Code. Cantidad unitSize
16416057 5 mg 5 miligramos
1 options
This item is not returnable. View return policy
Product Code. 16416057 Supplier MedChemExpress Supplier No. HY101297
This item is not returnable. View return policy
MedChemExpress Z-IETD-FMK (Z-IE(OMe)TD(OMe)-FMK) is a selective and cell permeable caspase-8 inhibitor. Z-IETD-FMK is also a granzyme B inhibitor.
Specifications
Pureza 98.0%
CAS 210344-98-2
Contenido y almacenamiento Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Para utilizar con (aplicación) Cancer-programmed cell death
Peso molecular 654.68 g/mol
Tipo de producto Peptides
Cantidad 5 mg
Condiciones de envío Room temperature
Solubilidad DMSO : 41.67 mg/mL (63.65 mM∣Need ultrasonic)
Sinónimo Z-IE(OMe)TD(OMe)-FMK