Learn More
YL-109, MedChemExpress
Código de producto. 16483557
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16483557 50 mg 50 miligramos
16467538 10 mg 10 miligramos
16450018 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16483557 Proveedor Medchem Express N.º de proveedor HY18619
MedChemExpress YL-109 is an antitumor agent that can induce carboxyl terminus of Hsp70-interacting protein (CHIP) expression through aryl hydrocarbon receptor (AhR) signaling. YL-109 has ability to inhibit breast cancer cell growth and invasiveness.
YL-109 is an antitumor agent that can induce carboxyl terminus of Hsp70-interacting protein (CHIP) expression through aryl hydrocarbon receptor (AhR) signaling. YL-109 has ability to inhibit breast cancer cell growth and invasiveness.
Especificaciones
Nombre del producto químico o material YL-109
CAS 36341-25-0
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C14H11NO2S
Cantidad 50 mg
Información de solubilidad DMSO : 100 mg/mL (388.64 mM; Need ultrasonic)
SMILES OC1=CC=C(C2=NC3=CC=CC=C3S2)C=C1OC
Peso molecular (g/mol) 257.31
Formula Weight (peso de la fórmula) 257.31
Porcentaje de pureza 98.74%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death