Learn More
Tocris Bioscience™ YKL 5-124
Descripción
YKL-5-124 is a potent and selective CDK7/Mat1/CycH and cyclin dependent kinase (CDK7) inhibitor (IC50 values = 9.7 nM and 53.5 nM, respectively). YKL-5-124 shows 100-fold selectivity over CDK9 and CDK2. YKL-5-124 induces cell cycle arrest at the G1/S transition, inhibits E2F-driven gene expression and inhibits CDK1 and CDK3 T-loop phosphorylation.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-[(1 S)-2-(Dimethylamino)-1-phenylethyl]-4,6-dihydro-6,6-dimethyl-3-[[4-[(1-oxo-2-propen-1-yl)amino]benzoyl]amino]pyrrolo[3,4-c]pyrazole-5(1 H)-carboxamide |
| CAS | 1957203-01-8 |
| Cantidad | 5 mg |
| Objetivo | Cyclin-dependent Kinase Inhibitors |
| Fórmula molecular | C28H33N7O3 |
| Pureza | 0.95 |