missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Yellow Fever Virus NS3 Polyclonal Antibody for Western Blot
Yellow fever virus (YFV) is an RNA virus belonging to the genus Flavivirus in the family Flaviviridae. YFV measures 40-60 nm in diameter and has a spherical, enveloped structure containing a single-stranded positive-sense RNA genome. The genome encodes three structural proteins (C, prM/M, and E) and seven nonstructural proteins (NS1, NS2A, NS2B, NS3, NS4A, NS4B, and NS5). YFV is transmitted by Aedes mosquitoes and is endemic in tropical regions of Africa and South America. It causes yellow fever, a hemorrhagic disease characterized by symptoms such as fever, jaundice, hemorrhage, and organ failure. Pathogenesis involves liver infection leading to hepatocyte and Kupffer cell damage, with the virus utilizing specific genes and molecular mechanisms for neurotropism and viscerotropism.
Especificaciones
Especificaciones
| Antígeno | Yellow Fever Virus NS3 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1.19 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Especie del huésped | Rabbit |
| Inmunógeno | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Yellow Fever Virus NS3 Protein (Yellow fever virus (strain 17D vaccine)). The exact sequence is proprietary. |
| Método de purificación | Antigen affinity chromatography |
| Cantidad | 100 μL |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
