Learn More
Y-27632, MedChemExpress
Descripción
Y-27632 attenuates Doxorubicin-induced apoptosis of human cardiac stem cells. Y-27632 also suppresses dissociation-induced apoptosis of murine prostate stem/progenitor cells. Y-27632 primes human induced pluripotent stem cells (hIPSCs) to selectively differentiate towards mesendodermal lineage via epithelial-mesenchymal transition-like modulation.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Y-27632 |
| CAS | 146986-50-7 |
| Color | Off-White |
| Notas de grado de pureza | Research |
| Fórmula molecular | C14H21N3O |
| Cantidad | 5 mg |
| Información de solubilidad | DMSO : 50 mg/mL (202.15 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.04 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C([C@@H]1CC[C@]([C@H](N)C)([H])CC1)NC2=CC=NC=C2 |
| Peso molecular (g/mol) | 247.34 |
| Formula Weight (peso de la fórmula) | 247.34 |
| Mostrar más |